Learn More
Agilent Technologies XF Loading Guide, For XFe96 and XF Pro, 6/Pk.
Catalog No. NC3449748
Encompass
The Seahorse XF Loading Guide is designed to streamline assay workflows by ensuring accurate port loading on XFe96 and XF Pro cartridges. By providing precise alignment, this tool helps reduce data outliers caused by inconsistent port loading, facilitating the acquisition of high-quality XF data.
  • Ensures precise loading of designated ports on XFe96 and XF Pro cartridges
  • Reduces data outliers caused by port loading errors
  • Includes 6 pairs of loading guides
Catalog No. NC3449748 Supplier Agilent Technologies Supplier No. 103786100
The Seahorse XF Loading Guide is designed to streamline assay workflows by ensuring accurate port loading on XFe96 and XF Pro cartridges. By providing precise alignment, this tool helps reduce data outliers caused by inconsistent port loading, facilitating the acquisition of high-quality XF data.
  • Ensures precise loading of designated ports on XFe96 and XF Pro cartridges
  • Reduces data outliers caused by port loading errors
  • Includes 6 pairs of loading guides