Learn More
Agilent Technologies Seahorse XF Pro M FluxPak. For use with XF Pro Analyzer.
Catalog No. NC2855237
Encompass
Seahorse XF Pro M FluxPak. For use with XF Pro Analyzer. Seahorse XF Pro M FluxPak includes 18 XFe96 sensor cartridges, 18 XF Pro M Cell Culture Microplates, and 1 bottle of Seahorse XF Calibrant Solution 500mL.
Catalog No. NC2855237 Supplier Agilent Technologies Supplier No. 103775100
Seahorse XF Pro M FluxPak. For use with XF Pro Analyzer. Seahorse XF Pro M FluxPak includes 18 XFe96 sensor cartridges, 18 XF Pro M Cell Culture Microplates, and 1 bottle of Seahorse XF Calibrant Solution 500mL.