Learn More
Agilent Technologies Seahorse XF Real-Time ATP Rate Assay Kit
Catalog No. NC1768680
Encompass
Each kit contains six (6) single-use pouches. Each pouch contains one (1) each of oligomycin and rotenone/antimycin A. For use with Seahorse XFe/XF96 and XFe24 analyzers.
Catalog No. NC1768680 Supplier Agilent Technologies Supplier No. 103592100
Each kit contains six (6) single-use pouches. Each pouch contains one (1) each of oligomycin and rotenone/antimycin A. For use with Seahorse XFe/XF96 and XFe24 analyzers.