Learn More
Agilent Technologies SEAHORSE XF T CELL META KIT
Catalog No. NC2145901
Encompass
Seahorse XF T Cell Metabolic Profiling KitSeahorse XF T Cell Metabolic Profiling Kit. Each kit contains six (6) single-use pouches. Each pouch contains one (1) each of oligomycin A, BAM15, and rotenone/antimycin A. For use with Seahorse XF Pro and XFe96 analyzers.Assay Wells 6 x 96-wellAssays per Kit: 6Shipping Condition: Ambient TemperatureStorage Temp Conditions: Room temperature
This item is not returnable. View return policy
Catalog No. NC2145901 Supplier Agilent Technologies Supplier No. 103772100
Add to Cart
Add to Cart
Seahorse XF T Cell Metabolic Profiling KitSeahorse XF T Cell Metabolic Profiling Kit. Each kit contains six (6) single-use pouches. Each pouch contains one (1) each of oligomycin A, BAM15, and rotenone/antimycin A. For use with Seahorse XF Pro and XFe96 analyzers.Assay Wells 6 x 96-wellAssays per Kit: 6Shipping Condition: Ambient TemperatureStorage Temp Conditions: Room temperature