Learn More
Agilent Technologies Seahorse XFe96/XF Pro FluxPak Mini, 6 Sensor Cartridges, 6 Cell Culture Microplates, 500 mL Calibrant Solution, 96 Wells per microplate, Room Temperature Storage (4 to 30 °C)
Catalog No. NC2393853
Encompass
Seahorse XFe96/XF Pro FluxPak Mini, 6 Sensor Cartridges, 6 Cell Culture Microplates, 500 mL Calibrant Solution, 96 Wells per microplate, Room Temperature Storage (4 to 30 °C)
Catalog No. NC2393853 Supplier Agilent Technologies Supplier No. 103793100
Seahorse XFe96/XF Pro FluxPak Mini, 6 Sensor Cartridges, 6 Cell Culture Microplates, 500 mL Calibrant Solution, 96 Wells per microplate, Room Temperature Storage (4 to 30 °C)