Learn More
Agilent Technologies Seahorse XFe96/XF Pro FluxPak
Catalog No. NC2151382
Encompass
Seahorse XFe96/XF Pro FluxPak . For use with XFe96 and XF Pro Analyzer. Seahorse XFe96/XF Pro FluxPak includes 18 XFe96/XF Pro sensor cartridges,18 XFe96/XF Pro Cell Culture Microplates, and 1 bottle of Seahorse XF Calibrant Solution 500mL.
This item is not returnable. View return policy
Catalog No. NC2151382 Supplier Agilent Technologies Supplier No. 103792100
Seahorse XFe96/XF Pro FluxPak . For use with XFe96 and XF Pro Analyzer. Seahorse XFe96/XF Pro FluxPak includes 18 XFe96/XF Pro sensor cartridges,18 XFe96/XF Pro Cell Culture Microplates, and 1 bottle of Seahorse XF Calibrant Solution 500mL.