Learn More
Agilent Technologies Cell Culture Microplates, 96-Well, Tissue Culture Treated, 6/Pk.
Catalog No. NC3104853
Encompass
These microplates are designed for use with Seahorse XFe96 and XF Pro Analyzers to support reliable metabolic assays. Each plate is tissue culture treated to promote optimal adherence and growth of mammalian cell types including monolayers, suspension cells, and isolated mitochondria.
  • Tissue culture treated surface for optimum cell adherence and growth.
  • Conforms to ANSI/SLAS microplate standards for footprint dimensions and well positions.
  • Integrated molded stops prevent sensor cartridge sleeves from disrupting the cell monolayer.
  • Supplied in individual gamma-irradiated containers to ensure sterility.
  • Supports a wide range of configurations including 2D monolayers, 3D spheroids, and suspension cells.
  • Compatible with both XFe96 and XF Pro analyzer systems.
Catalog No. NC3104853 Supplier Agilent Technologies Supplier No. 103794100
These microplates are designed for use with Seahorse XFe96 and XF Pro Analyzers to support reliable metabolic assays. Each plate is tissue culture treated to promote optimal adherence and growth of mammalian cell types including monolayers, suspension cells, and isolated mitochondria.
  • Tissue culture treated surface for optimum cell adherence and growth.
  • Conforms to ANSI/SLAS microplate standards for footprint dimensions and well positions.
  • Integrated molded stops prevent sensor cartridge sleeves from disrupting the cell monolayer.
  • Supplied in individual gamma-irradiated containers to ensure sterility.
  • Supports a wide range of configurations including 2D monolayers, 3D spheroids, and suspension cells.
  • Compatible with both XFe96 and XF Pro analyzer systems.