Learn More
Lonza Walkersville SeaKem LE Agarose
Catalog No. 501132005
Encompass
SeaKem LE Agarose was the first and is still the world’s most trusted agarose for nucleic acid electrophoresis. Ideal for routine analysis of nucleic acids by gel electrophoresis and blotting, each SeaKem LE Gel sharply resolves DNA and provides consistent resolution from lot-to-lot. This molecular biology grade agarose has no detectable DNase or RNase activity and forms strong gels with low background upon ethidium bromide, SYBR Green, or GelStar Staining. Due to its low EEO, DNA will have a high electrophoretic mobility. SeaKem LE Agarose is also recommended for protein gel electrophoresis in applications such as Ouchterlony and radial immunodiffusion (RID).Benefits:Wide resolution range (100 bp - 23 kb)All Lonza Agarose is quality tested to certify performanceHigh gel strengthLow EEOContent:SeaKem LE Agarose 1 kg
Catalog No. 50-113-2005 Supplier Lonza Walkersville Supplier No. 50005
SeaKem LE Agarose was the first and is still the world’s most trusted agarose for nucleic acid electrophoresis. Ideal for routine analysis of nucleic acids by gel electrophoresis and blotting, each SeaKem LE Gel sharply resolves DNA and provides consistent resolution from lot-to-lot. This molecular biology grade agarose has no detectable DNase or RNase activity and forms strong gels with low background upon ethidium bromide, SYBR Green, or GelStar Staining. Due to its low EEO, DNA will have a high electrophoretic mobility. SeaKem LE Agarose is also recommended for protein gel electrophoresis in applications such as Ouchterlony and radial immunodiffusion (RID).Benefits:Wide resolution range (100 bp - 23 kb)All Lonza Agarose is quality tested to certify performanceHigh gel strengthLow EEOContent:SeaKem LE Agarose 1 kg