Learn More
MSC ZEP Floor Sealer, Water-Based, Multi-Surface, 5 gal
Catalog No. 502818440
Encompass
Zep Seal 25 Floor Sealer is a water-based product designed for sealing a variety of flooring materials. It provides effective protection for multiple surfaces, making it suitable for diverse applications.
  • Water-based composition
  • Suitable for ceramic tiles, marble, slate, terrazzo, concrete, and brick
  • Supplied in a 5 gal pail
  • White, opaque liquid with a mild odor and a pH of 8.02
  • Soluble in water
  • Not classified as a hazardous substance or mixture
Catalog No. 50-281-8440 Supplier MSC Supplier No. 17690447
Zep Seal 25 Floor Sealer is a water-based product designed for sealing a variety of flooring materials. It provides effective protection for multiple surfaces, making it suitable for diverse applications.
  • Water-based composition
  • Suitable for ceramic tiles, marble, slate, terrazzo, concrete, and brick
  • Supplied in a 5 gal pail
  • White, opaque liquid with a mild odor and a pH of 8.02
  • Soluble in water
  • Not classified as a hazardous substance or mixture