Learn More
Chemglass Life Sciences Crimp Seal, Black, Viton
SDP
Supplier:  Chemglass Life Sciences CV34500008
Encompass_Preferred
Crimp Seal, Black, Viton. Recommended for chlorinated solvents. Excellent chemical resistance.
Catalog No. 50-153-2551