Learn More
Waters Corp Plunger Seal, Clear-100
Supplier:  Waters Corp WAT022934
Encompass_Preferred
Replacement Plunger Seal. Made from a proprietary material, the Clear-100™ Plunger Seals feature an internal spring that provides the correct pressure on the plunger, increasing the life of the sapphire and the seal itself. The Clear-100™ Seal features an exclusive ringed or ribbed design on the inside of the seal, which helps to maintain a snug fit and increases the seal’s durability. The Clear-100™ Seal is best suited to most applications for Waters Solvent Delivery Systems. It is also available as a 4/pk via WAT022946. It is also included in Performance Maintenance Kits for 515, 1515, 1525 and 600 Pumps.
Catalog No. 50-819-111