Learn More
Waters Corp Waters Clear-100 Plunger Seals, 4/pk, HPLC
Catalog No. 50819011 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Clear-100™ Plunger Seals are crafted from a proprietary material, incorporating an internal spring to ensure optimal pressure on the plunger. This design enhances the longevity of both the sapphire and the seal.
  • Features an exclusive ringed or ribbed design on the inside, promoting a snug fit.
  • Increases the seal's durability.
  • Suitable for most applications in solvent delivery systems.
  • Included in performance maintenance kits.
Catalog No. 50-819-011 Supplier Waters Corp Supplier No. WAT022946
The Clear-100™ Plunger Seals are crafted from a proprietary material, incorporating an internal spring to ensure optimal pressure on the plunger. This design enhances the longevity of both the sapphire and the seal.
  • Features an exclusive ringed or ribbed design on the inside, promoting a snug fit.
  • Increases the seal's durability.
  • Suitable for most applications in solvent delivery systems.
  • Included in performance maintenance kits.