Learn More
Waters Corp Waters Seal Extension Tube, 0.004 in. ID, ACQUITY H-Class FTN System
Catalog No. 50793078 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Seal Extension Tube is designed for use with ACQUITY, ACQUITY Premier, and ACQUITY UPLC systems. It features a 0.004 in. ID and is specifically compatible with the ACQUITY UPLC H-Class System and ACQUITY Premier System.
  • Designed for use with UPLC systems
  • Compatible with H-Class System and Premier System models
  • Features a 0.004 in. ID
  • Categorized as a service part/kit
  • One unit per package
  • Type: Needle, Tubing
  • Replacement as required
Catalog No. 50-793-078 Supplier Waters Corp Supplier No. 700005234
This Seal Extension Tube is designed for use with ACQUITY, ACQUITY Premier, and ACQUITY UPLC systems. It features a 0.004 in. ID and is specifically compatible with the ACQUITY UPLC H-Class System and ACQUITY Premier System.
  • Designed for use with UPLC systems
  • Compatible with H-Class System and Premier System models
  • Features a 0.004 in. ID
  • Categorized as a service part/kit
  • One unit per package
  • Type: Needle, Tubing
  • Replacement as required