Learn More
BURKLE INC Seal-it security seal, red, LxW 7.01x1.18 in. (178x30 mm)
Catalog No. 5000025657
Encompass_Preferred
Retention samples are kept for later comparison. Clear proof that a retention sample matches the original requires marked, sealed containers—often impossible without eyelets.
seal-it solves this! The self-adhesive label sticks to all standard containers with strong, tamper-evident adhesion.
Once sealed, the container cannot be opened without destroying the seal-it label. Its cross-perforations tear instantly if tampered, like a motor vehicle tax disc.
seal-it securely seals bottles, sample bags, sacks, envelopes, and more. The aluminum foil adapts to any shape, and the white label area is writable with all common pens for content, date, operator name, and more.
Catalog No. 50-000-25657 Supplier BURKLE INC Supplier No. 53990003
Retention samples are kept for later comparison. Clear proof that a retention sample matches the original requires marked, sealed containers—often impossible without eyelets.
seal-it solves this! The self-adhesive label sticks to all standard containers with strong, tamper-evident adhesion.
Once sealed, the container cannot be opened without destroying the seal-it label. Its cross-perforations tear instantly if tampered, like a motor vehicle tax disc.
seal-it securely seals bottles, sample bags, sacks, envelopes, and more. The aluminum foil adapts to any shape, and the white label area is writable with all common pens for content, date, operator name, and more.