Learn More
Waters Corp WATERS CORPORATION Piston Seal Kit, Rebuild Kit, HPLC
Catalog No. 50818894 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Designed specifically for the Reagent Manager, the Piston Seal Kit features an internal spring that provides the correct pressure on the plunger, increasing the life of the piston.
  • Contains one piston seal.
  • Includes one backup washer.
  • Provides one seal wash diaphragm.
  • Comes with one seal insertion/removal tool.
  • Includes one scouring pad to clean the piston when changing the seal.
Catalog No. 50-818-894 Supplier Waters Corp Supplier No. 700000152
Designed specifically for the Reagent Manager, the Piston Seal Kit features an internal spring that provides the correct pressure on the plunger, increasing the life of the piston.
  • Contains one piston seal.
  • Includes one backup washer.
  • Provides one seal wash diaphragm.
  • Comes with one seal insertion/removal tool.
  • Includes one scouring pad to clean the piston when changing the seal.