Learn More
*KLAREX HEALTH LLC Seal’ N Freeze Box
Catalog No. NC1877501
Encompass_Preferred
The Seal’N Freeze Box allows users to freeze up to four tissue samples simultaneously and hands-free, when used with Seal’N Freeze Cryotray cryomolds inserts. The box’s stainless-steel case and foam lining ensures optimal temperature control throughout the hands-free freezing process; and its small and compact size ensures easy storage – even in the most cluttered laboratory. See how it works: https://www.youtube.com/watch?v=3sMTXM32vjg.
Catalog No. NC1877501 Supplier *KLAREX HEALTH LLC Supplier No. 1000
The Seal’N Freeze Box allows users to freeze up to four tissue samples simultaneously and hands-free, when used with Seal’N Freeze Cryotray cryomolds inserts. The box’s stainless-steel case and foam lining ensures optimal temperature control throughout the hands-free freezing process; and its small and compact size ensures easy storage – even in the most cluttered laboratory. See how it works: https://www.youtube.com/watch?v=3sMTXM32vjg.