Learn More
*KLAREX HEALTH LLC Seal’ N Freeze Cryotray Large cryomolds, 200 molds
Catalog No. NC1877500
Encompass_Preferred
Seal’N Freeze Cryotray cryomolds are compact disposable plastic embedding molds with attached sealable lids that allow for batch processing of tissue samples in perfect uniform blocks that are ideal for cryosectioning and storage. Each disposable plastic Seal’N Freeze Cryotray includes four individual disposable cryomolds which can be easily separated from the center connector by folding and tearing. Maximize your efficiency by using the Cryotray embedding molds with our compact Seal’N Freeze Box, which allows for convenient hands-free freezing. See how it works: https://www.youtube.com/watch?v=3sMTXM32vjg 25 x 30 x 20 mm (LxWxD) 50 pack of Cryotrays (200 base cryomolds)
Catalog No. NC1877500 Supplier *KLAREX HEALTH LLC Supplier No. 1002
Seal’N Freeze Cryotray cryomolds are compact disposable plastic embedding molds with attached sealable lids that allow for batch processing of tissue samples in perfect uniform blocks that are ideal for cryosectioning and storage. Each disposable plastic Seal’N Freeze Cryotray includes four individual disposable cryomolds which can be easily separated from the center connector by folding and tearing. Maximize your efficiency by using the Cryotray embedding molds with our compact Seal’N Freeze Box, which allows for convenient hands-free freezing. See how it works: https://www.youtube.com/watch?v=3sMTXM32vjg 25 x 30 x 20 mm (LxWxD) 50 pack of Cryotrays (200 base cryomolds)