Learn More
Waters Corp Waters Seal Pack and Needle Replacement Kit, Rebuild Kit, HPLC, 1 pk
Catalog No. 501534810 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Seal Pack and Needle Replacement Kit includes one stainless steel annealed ferrule, one short compression screw (for old style injector only), one needle, one long compression screw (for new style injector only), one seal pak assembly, and the installation instructions.
  • Contains all necessary parts for replacement
  • Includes components for both old and new style injectors (short and long compression screws)
Catalog No. 50-153-4810 Supplier Waters Corp Supplier No. WAT045559
The Seal Pack and Needle Replacement Kit includes one stainless steel annealed ferrule, one short compression screw (for old style injector only), one needle, one long compression screw (for new style injector only), one seal pak assembly, and the installation instructions.
  • Contains all necessary parts for replacement
  • Includes components for both old and new style injectors (short and long compression screws)