Learn More
Andwin Scientific SealPlate Plate Sealing Film, 100 per case
SDP
Catalog No. NC9479592
Encompass_Preferred
The SealPlate Sealing Films allows the reduction of evaporation, spillage, and contamination between wells. The film offers secure seal by removing the risks of "edge effects" in sensitive ELISA assays. This product can also be utilized for short-term storage, incubation, and containment of biohazards. It is non-pierceable and is measured to have a dimension of 79.4 x 146.1 mm. This product can work within the temperature range of -40C to +120C.
Catalog No. NC9479592 Supplier Andwin Scientific Supplier No. 100SEALPLT
The SealPlate Sealing Films allows the reduction of evaporation, spillage, and contamination between wells. The film offers secure seal by removing the risks of "edge effects" in sensitive ELISA assays. This product can also be utilized for short-term storage, incubation, and containment of biohazards. It is non-pierceable and is measured to have a dimension of 79.4 x 146.1 mm. This product can work within the temperature range of -40C to +120C.