Learn More
Perkin Elmer US LLC Seal Plug for Impact Bead of AAnalyst AA Instruments
Catalog No. 502117238
Encompass_Preferred
Replacement seal plug for the 00572615 Pyrex impact bead used with end caps of metal body nebulizers on AAnalyst Atomic Absorption (AA) instruments.
Catalog No. 50-211-7238 Supplier Perkin Elmer US LLC Supplier No. 00572624
Replacement seal plug for the 00572615 Pyrex impact bead used with end caps of metal body nebulizers on AAnalyst Atomic Absorption (AA) instruments.