Learn More
Globe Scientific Caps for Automation
Click to view available options
For Use With (Equipment):
Beckman DXA5000
Roche E801
Description
- Globe Scientific caps for automated systems fit several of the most popular platforms and fit seamlessly into capping processes.
- These cost-effective alternatives to OEM caps are manufactured at high standards to ensure efficient sample processing.
- Seamless cost-effective replacements for OEM caps
- Smooth operation in existing platforms
- Reliable sealing during high-speed transport and agitation
- Made from high-grade Polyethylene (PE) material
- Non-Sterile
Specifications
Specifications
Type | Automated Sampling System Cap |
For Use With (Equipment) | Roche E801 |
Note: All instruments and names of manufacturers mentioned herein are the trademarks of their respective companies and are used for reference purposes only. Globe Scientific LLC. Expressly disclaims any affiliation to them