Learn More
Perkin Elmer US LLC Seal Removal Tool
Catalog No. 502113231
Encompass_Preferred
Calion standards are premixed and adsorbed into a solid phase. These standards are ideal for in field use because they contain no liquids. Calion PV Mixes are used for Torion T-9 calibration, including both mass and GC retention time calibrations. The combination of Custodion SPME sampling with Calion standards provides robust and rapid capability for in-field calibration because they contain no liquids. Calion PV Mixes are used for Torion T-9 calibration, including both mass and GC retention time calibrations. The combination of Custodion SPME sampling with Calion standards provides robust and rapid capability for in-field calibration.
Catalog No. 50-211-3231 Supplier Perkin Elmer US LLC Supplier No. NTSM0060124
Calion standards are premixed and adsorbed into a solid phase. These standards are ideal for in field use because they contain no liquids. Calion PV Mixes are used for Torion T-9 calibration, including both mass and GC retention time calibrations. The combination of Custodion SPME sampling with Calion standards provides robust and rapid capability for in-field calibration because they contain no liquids. Calion PV Mixes are used for Torion T-9 calibration, including both mass and GC retention time calibrations. The combination of Custodion SPME sampling with Calion standards provides robust and rapid capability for in-field calibration.