Learn More
Waters Corp Waters Quality Parts Plunger Seal, GFP, Black, HPLC
Catalog No. 50818751 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Made from high-performance, glass-filled polymer, the Plunger Seal features an internal spring designed to provide the correct pressure on the plunger, extending the life of both the sapphire plunger and the seal. While the Clear-100™ Seal is typically used in solvent delivery systems and Performance Maintenance Kits, the GFP seal may be more suitable for certain applications.
  • Made from GFP (glass-filled polymer).
  • Black color.
  • Designed for HPLC system types.
  • Includes 1 unit per package.
  • Replacement is on an as-required basis.
Catalog No. 50-818-751 Supplier Waters Corp Supplier No. WAT026613
Made from high-performance, glass-filled polymer, the Plunger Seal features an internal spring designed to provide the correct pressure on the plunger, extending the life of both the sapphire plunger and the seal. While the Clear-100™ Seal is typically used in solvent delivery systems and Performance Maintenance Kits, the GFP seal may be more suitable for certain applications.
  • Made from GFP (glass-filled polymer).
  • Black color.
  • Designed for HPLC system types.
  • Includes 1 unit per package.
  • Replacement is on an as-required basis.