Learn More
BUCHI Seal with Ring Support
SDP
Catalog No. 05405663 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Equipment):
KjelMaster K-375
Catalog No. For Use With (Equipment)
05-405-663 KjelMaster K-375
1 options
Catalog No. 05-405-663 Supplier BUCHI Supplier No. 11057039
Add to Cart
Add to Cart
Support for seal with ring, a screw threaded connection piece to complete sample tube connection on BUCHI KjelMaster K-375.
  • Holder for the seal with plastic ring on Kjeldahl Distillation Units
Specifications
For Use With (Application) Sample Tube Connection
Type Support
Material Plastic
For Use With (Equipment) KjelMaster K-375