Learn More
Agilent Technologies Plate Seal, 96-Well, Solid, Santoprene, 2 mL, 100/Pk.
Catalog No. 50995892
Encompass
This solid, 96-well plate seal mat is constructed from Santoprene, a durable thermoplastic elastomer, and is specifically engineered to accommodate 2 mL well storage plate volumes. The design provides a secure fit to maintain sample integrity and is compatible with automated systems.
  • Designed for 96-well storage plates with 2 mL capacities
  • Constructed from chemical-resistant thermoplastic elastomer material
  • Robot-friendly design
  • Low permeability to alcohols
  • Pack size of 100 per unit
Catalog No. 50-995-892 Supplier Agilent Technologies Supplier No. 201160100
This solid, 96-well plate seal mat is constructed from Santoprene, a durable thermoplastic elastomer, and is specifically engineered to accommodate 2 mL well storage plate volumes. The design provides a secure fit to maintain sample integrity and is compatible with automated systems.
  • Designed for 96-well storage plates with 2 mL capacities
  • Constructed from chemical-resistant thermoplastic elastomer material
  • Robot-friendly design
  • Low permeability to alcohols
  • Pack size of 100 per unit