Learn More
Agilent Technologies Plate Seal and Mat, 96-Well, Santoprene, For 2 mL Fritted Filter Plate, 100/Pk.
Catalog No. 50995893
Encompass
These plate seals and mats are designed to provide both top and bottom coverage for 96-well configurations, ensuring optimal sample protection and processing. Manufactured from high-quality Santoprene, these seals are specifically engineered for use with 2 mL fritted filter plates.
  • Designed for 96-well plate configurations
  • Provides both top and bottom sealing surfaces
  • Constructed from durable Santoprene
  • Compatible with 2 mL fritted filter plates
  • Supplied in a 100/pk quantity
Catalog No. 50-995-893 Supplier Agilent Technologies Supplier No. 201162100
These plate seals and mats are designed to provide both top and bottom coverage for 96-well configurations, ensuring optimal sample protection and processing. Manufactured from high-quality Santoprene, these seals are specifically engineered for use with 2 mL fritted filter plates.
  • Designed for 96-well plate configurations
  • Provides both top and bottom sealing surfaces
  • Constructed from durable Santoprene
  • Compatible with 2 mL fritted filter plates
  • Supplied in a 100/pk quantity