Learn More
Chemglass Life Sciences Aluminum Seal, Pressure Release, Silver, 20mm, PTFE/Gray Butyl Rubber Septa
SDP
Catalog No. 501532537
Encompass_Preferred
Aluminum Seal, Pressure Release, Silver, 20mm, PTFE/Gray Butyl Rubber Septa. Pressure release seals are designed to open when the internal pressure exceeds 3.00.5 bar.
Catalog No. 50-153-2537 Supplier Chemglass Life Sciences Supplier No. CV32010020
Aluminum Seal, Pressure Release, Silver, 20mm, PTFE/Gray Butyl Rubber Septa. Pressure release seals are designed to open when the internal pressure exceeds 3.00.5 bar.