Learn More
Chemglass Life Sciences Crimp Seal, Silver, 10mil PTFE
SDP
Catalog No. 501532524
Encompass_Preferred
Crimp Seal Selection Guide:PTFE/Red Rubber -Medium durometer red rubber with a .015” layer of clear PTFE. Great and economical for routine analysis. Not recommended for multiple injections.PTFE/Silicone -Pure Silicone laminated with .005” of PTFE. Recommended when multiple injections are required. Resist coring and excellent resealing qualities.PTFE/Silicone/PTFE -Medium durometer silicone with .003” silicone laminated on both sides. Great for trace analysis work. Resist coring and good resealing characteristics. Use with auto samplers with blunt tip syringes.Viton -Recommended for chlorinated solvents. Excellent chemical resistance.Polypropylene -Not resealable. For one time use only. Easily penetrated by syringes and chemical resistant.
Catalog No. 50-153-2524 Supplier Chemglass Life Sciences Supplier No. CV31000008
Crimp Seal Selection Guide:PTFE/Red Rubber -Medium durometer red rubber with a .015” layer of clear PTFE. Great and economical for routine analysis. Not recommended for multiple injections.PTFE/Silicone -Pure Silicone laminated with .005” of PTFE. Recommended when multiple injections are required. Resist coring and excellent resealing qualities.PTFE/Silicone/PTFE -Medium durometer silicone with .003” silicone laminated on both sides. Great for trace analysis work. Resist coring and good resealing characteristics. Use with auto samplers with blunt tip syringes.Viton -Recommended for chlorinated solvents. Excellent chemical resistance.Polypropylene -Not resealable. For one time use only. Easily penetrated by syringes and chemical resistant.