Learn More
Chemglass Life Sciences Crimp Seal, Silver, Clear PTFE/Red Rubber
SDP
Catalog No. 501532526
Encompass_Preferred
Crimp Seal, Silver, Clear PTFE/Red Rubber. Medium durometer red rubber with a .015 layer of clear PTFE. Great and economical for routine analysis. Not recommended for multiple injections.
Catalog No. 50-153-2526 Supplier Chemglass Life Sciences Supplier No. CV31100008
Crimp Seal, Silver, Clear PTFE/Red Rubber. Medium durometer red rubber with a .015 layer of clear PTFE. Great and economical for routine analysis. Not recommended for multiple injections.