Learn More
Electron Microscopy Sciences ProPlate™ MicroArray Slide Seal Strips and Applicator
SDP
Catalog No. 5034303
Encompass_Preferred
The plastic applicator helps to apply seal strips to the top surface of the chambers. Seal strips are ideal to prevent evaporation, they remove cleanly and easily without leaving a residue.
Catalog No. 50-343-03 Supplier Electron Microscopy Sciences Supplier No. 6348480
The plastic applicator helps to apply seal strips to the top surface of the chambers. Seal strips are ideal to prevent evaporation, they remove cleanly and easily without leaving a residue.