Learn More
Electron Microscopy Sciences Flexwell™ Seal Strips
SDP
Catalog No. 5034595
Encompass_Preferred
Use with FlexWell removable incubation chambers. 24mm X 77mm seal area..
Catalog No. 50-345-95 Supplier Electron Microscopy Sciences Supplier No. 6348553
Use with FlexWell removable incubation chambers. 24mm X 77mm seal area..