Learn More
Waters Corp Waters Seal Wash Face Seal, PTFE, HPLC System
Catalog No. 502039979 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These seals are an integral part of the seal wash system within the 2695 Separations Modules, designed to maximize plunger seal life in HPLC systems.
  • Integral part of the seal wash system
  • Maximizes plunger seal life
  • Compatible with 2695 Separations Module and 2695 D Dissolution Separations Module
  • Manufactured from PTFE
  • Annual replacement schedule
  • Provided as one unit per package
Catalog No. 50-203-9979 Supplier Waters Corp Supplier No. WAT271017
These seals are an integral part of the seal wash system within the 2695 Separations Modules, designed to maximize plunger seal life in HPLC systems.
  • Integral part of the seal wash system
  • Maximizes plunger seal life
  • Compatible with 2695 Separations Module and 2695 D Dissolution Separations Module
  • Manufactured from PTFE
  • Annual replacement schedule
  • Provided as one unit per package