Learn More
Waters Corp WATERS CORPORATION Seal Wash Housing, Replacement, 2525 Binary Gradient Module, Assembly
Catalog No. 50792596 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This seal wash housing is a replacement part for a 2525 Binary Gradient Module. It is designed for use in purification systems.
  • Replacement part for a 2525 binary gradient module.
  • Compatible with 2535 quaternary gradient manager.
  • Compatible with 2545 (Q) binary gradient manager.
  • Supplied as an assembly.
  • Designed for purification systems.
  • For use with UPC2 instruments.
Catalog No. 50-792-596 Supplier Waters Corp Supplier No. 700001531
This seal wash housing is a replacement part for a 2525 Binary Gradient Module. It is designed for use in purification systems.
  • Replacement part for a 2525 binary gradient module.
  • Compatible with 2535 quaternary gradient manager.
  • Compatible with 2545 (Q) binary gradient manager.
  • Supplied as an assembly.
  • Designed for purification systems.
  • For use with UPC2 instruments.