Learn More
Waters Corp Waters Seal Wash Housing, UPLC Pump, Stainless Steel
Catalog No. 50792773 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Seal Wash Housing is a replacement component designed for UPLC Pumps. It is compatible with various ACQUITY UPLC systems, including Binary Solvent Managers, Isocratic and Quaternary Solvent Managers, and NanoACQUITY Solvent Managers. It serves as a spare part for ACQUITY UPLC System and ACQUITY UPLC H-Class System.
  • Made from stainless steel
  • For UPLC system type
  • Compatible with ACQUITY UPLC ISM, ACQUITY UPLC QSM, ACQUITY UPLC H-Class QSM, nanoACQUITY ASM, nanoACQUITY BSM components
  • Instrument compatibility includes ACQUITY UPLC H-Class System, nanoACQUITY UPLC System, ACQUITY
  • Type: Pump head
Catalog No. 50-792-773 Supplier Waters Corp Supplier No. 700002597
This Seal Wash Housing is a replacement component designed for UPLC Pumps. It is compatible with various ACQUITY UPLC systems, including Binary Solvent Managers, Isocratic and Quaternary Solvent Managers, and NanoACQUITY Solvent Managers. It serves as a spare part for ACQUITY UPLC System and ACQUITY UPLC H-Class System.
  • Made from stainless steel
  • For UPLC system type
  • Compatible with ACQUITY UPLC ISM, ACQUITY UPLC QSM, ACQUITY UPLC H-Class QSM, nanoACQUITY ASM, nanoACQUITY BSM components
  • Instrument compatibility includes ACQUITY UPLC H-Class System, nanoACQUITY UPLC System, ACQUITY
  • Type: Pump head