Learn More
Waters Corp Waters Corporation Seal Wash Housing Seal, 0.0787 in. ID, Float Flanged, Pack of 2
Catalog No. 502039439 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement seal wash housing seal is designed for pump head assemblies. It is found in ACQUITY Solvent Managers, H-Class Quaternary Solvent Managers and NanoACQUITY Solvent Managers.
  • Internal diameter of 0.0787 in.
  • Float flanged
  • Supplied in a pack of 2
  • Recommended for annual replacement
  • Pump head type
  • UPLC system type
Catalog No. 50-203-9439 Supplier Waters Corp Supplier No. 700002598
This replacement seal wash housing seal is designed for pump head assemblies. It is found in ACQUITY Solvent Managers, H-Class Quaternary Solvent Managers and NanoACQUITY Solvent Managers.
  • Internal diameter of 0.0787 in.
  • Float flanged
  • Supplied in a pack of 2
  • Recommended for annual replacement
  • Pump head type
  • UPLC system type