Learn More
Waters Corp Waters Corporation Seal Wash Plunger Seal Replacement Kit, 2 pk, for Alliance HPLC Systems
Supplier:  Waters Corp WAT271018
Encompass_Preferred
The Seal Wash Plunger Seal Replacement Kit is designed for use with Alliance HPLC Systems, specifically the 2695 Separations Modules. This kit is an integral part of the seal wash system, helping to maximize plunger seal life and ensure optimal performance. It is recommended for annual replacement to maintain system integrity.
  • Integral to the 2695 Separations Modules
  • Helps ensure that plunger seal life is maximized
  • Recommended for annual replacement
  • Includes 2 units per package
  • For use with HPLC systems
Catalog No. 50-203-9980