Learn More
Waters Corp Waters Seal Wash Priming Adapter, Quality Part, 1 pk, for Alliance HPLC System
Catalog No. 50793808 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This seal wash priming adapter is a Waters Quality part designed for use with the Alliance HPLC system 2695 Separations Module and the 2695D Dissolution Separations Module.
  • Provides priming for seal wash.
  • Quality part.
  • Compatible with Alliance HPLC system 2695 Separations Module.
  • Compatible with 2695D Dissolution Separations Module.
  • Supplied as 1 unit per package.
  • Replaced as required.
Catalog No. 50-793-808 Supplier Waters Corp Supplier No. WAT270586
This seal wash priming adapter is a Waters Quality part designed for use with the Alliance HPLC system 2695 Separations Module and the 2695D Dissolution Separations Module.
  • Provides priming for seal wash.
  • Quality part.
  • Compatible with Alliance HPLC system 2695 Separations Module.
  • Compatible with 2695D Dissolution Separations Module.
  • Supplied as 1 unit per package.
  • Replaced as required.