Learn More
Med Vet International McKesson Capillary Tube Holding Tray, 24 Place, 553, 10/Box
Catalog No. 503043928
Encompass_Preferred
Capillary Tube Holding Tray McKesson 24 Place, Wax Sealant For Microhematocrit Capillary Tubes With 24 numbered places for easy patient identification Contains a wax compound that quickly forms a tight seal The sealant is formulated to withstand centrifugation There is no need for plastic caps or heat sealing 24 place, wax sealant
Catalog No. 50-304-3928 Supplier Med Vet International Supplier No. 862529BX
Capillary Tube Holding Tray McKesson 24 Place, Wax Sealant For Microhematocrit Capillary Tubes With 24 numbered places for easy patient identification Contains a wax compound that quickly forms a tight seal The sealant is formulated to withstand centrifugation There is no need for plastic caps or heat sealing 24 place, wax sealant