Learn More
Enterprise Technology Solutions Sealed Air Tuffgard Extreme Mailers, 7-1/4"x12", 50/CT, WE
SDP
Catalog No. 5000827312
Encompass_Preferred
No. 1 mailers are tough and durable to handle your most demanding mailing applications. Multilayer, co-extruded film resists moisture, punctures and tears for reliable containment. Barrier bubble layer offers excellent air retention and cushioning to keep your items protected. A seamless bottom increases its cushioning protection for enhanced security. Self-seal adhesive provides a quick and secure closure without requiring moisture.
Catalog No. 50-008-27312 Supplier Enterprise Technology Solutions Supplier No. SEL10080
No. 1 mailers are tough and durable to handle your most demanding mailing applications. Multilayer, co-extruded film resists moisture, punctures and tears for reliable containment. Barrier bubble layer offers excellent air retention and cushioning to keep your items protected. A seamless bottom increases its cushioning protection for enhanced security. Self-seal adhesive provides a quick and secure closure without requiring moisture.