Learn More
Bio Rad Laboratories SEALED ICE BLOCKS
Catalog No. 509011957
Encompass
Block, Ice; Bio-Rad; Sealed; For Criterion blotter; Absorb heat generated during rapid transfers; 2
Catalog No. 50-901-1957 Supplier Bio Rad Laboratories Supplier No. 1704087
Block, Ice; Bio-Rad; Sealed; For Criterion blotter; Absorb heat generated during rapid transfers; 2