Learn More
Grainger Sealed Lead Acid Battery 12VDC 18Ah
Catalog No. 502767173
Encompass_Preferred
The Sealed Lead Acid Battery 12V 16Ah from EnerSys is a maintenance free, valve regulated lead acid (VRLA) battery. It has a spill proof construction and is designed for deep cycle applications. This 12 volt battery has a capacity of 16 amp hours and measures 7.13 inches x 3.01 inches x 6.57 inches (LxWxH). It has stainless steel bolts and copper alloy terminals. The operating temperature range is -40°F to 160°F.
Catalog No. 50-276-7173 Supplier Grainger Supplier No. 16X249
The Sealed Lead Acid Battery 12V 16Ah from EnerSys is a maintenance free, valve regulated lead acid (VRLA) battery. It has a spill proof construction and is designed for deep cycle applications. This 12 volt battery has a capacity of 16 amp hours and measures 7.13 inches x 3.01 inches x 6.57 inches (LxWxH). It has stainless steel bolts and copper alloy terminals. The operating temperature range is -40°F to 160°F.