Learn More
Medicus Health Sealed Specimen Transport Tote, Clear
SDP
Catalog No. NC1236071
Encompass
∙ Sealed Specimen Transport Tote is great for transporting clinical and biological samples that you want to keep clean and dry ∙Translucent totes that allow the user to visually check for sample integrity∙Great for carrying blood tubes, specimen transport bags, urine cups and tube racks∙Totes are autoclavable. The unit should be unlocked and open during autoclaving process with silicone seal left in place∙Sealed Transport Tote is in compliance with OSHA 29CFR 1910 1030 ∙Also works for transporting chemo bags throughout the hospital∙Durable, translucent, polycarbonate construction∙Lid opens 180 degrees to allow for full access to the contents∙Each tote includes one long divider and two short dividers∙Exterior dimensions: 15"L x 7.75"W x 6.5"H; Interior dimensions: 13"L x 6.25"W x 5.25"H
Catalog No. NC1236071 Supplier Medicus Health Supplier No. 7018M1
∙ Sealed Specimen Transport Tote is great for transporting clinical and biological samples that you want to keep clean and dry ∙Translucent totes that allow the user to visually check for sample integrity∙Great for carrying blood tubes, specimen transport bags, urine cups and tube racks∙Totes are autoclavable. The unit should be unlocked and open during autoclaving process with silicone seal left in place∙Sealed Transport Tote is in compliance with OSHA 29CFR 1910 1030 ∙Also works for transporting chemo bags throughout the hospital∙Durable, translucent, polycarbonate construction∙Lid opens 180 degrees to allow for full access to the contents∙Each tote includes one long divider and two short dividers∙Exterior dimensions: 15"L x 7.75"W x 6.5"H; Interior dimensions: 13"L x 6.25"W x 5.25"H