Learn More
Malvern Panalytical Sealed Toluene Cuvette
Catalog No. NC2053930
Encompass
Toluene scattering intensity test sample. Analar grade Toluene  permanently sealed in 12mm square quartz fluorescence cuvette. Used for calibrating Zetasizer Advance Systems capable of Molecular Weight and MADLS Partilce concentration and for checking the scattering intensity of other systems with a size measurement capability. Compatible with all Zetasizer systems including: Zetasizer Adavance range, Zetasizer Nano series and Zetasizer µV.
Catalog No. NC2053930 Supplier Malvern Panalytical Supplier No. 10019464
Toluene scattering intensity test sample. Analar grade Toluene  permanently sealed in 12mm square quartz fluorescence cuvette. Used for calibrating Zetasizer Advance Systems capable of Molecular Weight and MADLS Partilce concentration and for checking the scattering intensity of other systems with a size measurement capability. Compatible with all Zetasizer systems including: Zetasizer Adavance range, Zetasizer Nano series and Zetasizer µV.