Learn More
Bolle Safety™ Sealed Universal Safety Goggles
Catalog No. 17467450
Change view
Click to view available options
Style:
Clear
Clear and Foam
Clear with Neoprene Strap
Smoke and Foam
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
17-467-450 Clear
17-467-445 Smoke and Foam
17-467-447 Clear and Foam
17-467-449 Clear with Neoprene Strap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-467-450 Supplier Bolle Safety™ Supplier No. UNIVGN11W
Add to Cart
Add to Cart
This goggle provides optimum protection and comfort in all circumstances thanks to its ergonomic design. This innovative goggle with its exclusive platinum coating is available in several versions, each adapted to a specific work environment to offer more peace of mind and efficiency for the worker.
  • Optimal fit thanks to its Wave technology
  • PC/TPR co-injected frame with overflow chute
  • Effortless pivoting faceguard
  • Improved recyclability
  • UV protection
  • Scratch-resistant
  • Anti-fog
Specifications
Lens Coating Scratch-resistant, Anti-fog
Description Sealed
Color Black and Green
Lens Tint Clear
Product Type Safety Goggles
Style Clear
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
Frame Type Standard