Learn More
MSC Value Collection Impulse Sealer Service Kit, 18 in, 1 ea
Catalog No. 501919400
Encompass
This impulse sealer service kit is designed for an 18 in Max Seal impulse sealer, enabling perfect seals across various poly thicknesses. It provides essential components for maintenance and repair.
  • Includes (1) PTFE tape strip
  • Includes (1) PTFE cover
  • Includes (2) heating elements
  • Manufactured in Taiwan
  • Designed for an 18 in maximum seal size
Catalog No. 50-191-9400 Supplier MSC Supplier No. 39590138
This impulse sealer service kit is designed for an 18 in Max Seal impulse sealer, enabling perfect seals across various poly thicknesses. It provides essential components for maintenance and repair.
  • Includes (1) PTFE tape strip
  • Includes (1) PTFE cover
  • Includes (2) heating elements
  • Manufactured in Taiwan
  • Designed for an 18 in maximum seal size