Learn More
Waters Corp Waters Sample Collection Plate Sealing Cap, PTFE/Silicone, Reusable, for 96- and 384-well Plates
Catalog No. 50818658 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This reusable cap mat is designed to protect samples from contaminants during preparation. Constructed from PTFE/silicone, it allows for easy cleaning with organic solvents and reuse. It securely seals 96- and 384-well plates, ensuring individual wells are protected from contaminants to maintain sample integrity.
  • Protects samples from contaminants.
  • PTFE/silicone construction.
  • Reusable and easy to clean with organic solvents.
  • Compatible with 96- and 384-well plates.
  • Ensures individual well sealing.
  • Maintains sample integrity.
Catalog No. 50-818-658 Supplier Waters Corp Supplier No. WAT058959
This reusable cap mat is designed to protect samples from contaminants during preparation. Constructed from PTFE/silicone, it allows for easy cleaning with organic solvents and reuse. It securely seals 96- and 384-well plates, ensuring individual wells are protected from contaminants to maintain sample integrity.
  • Protects samples from contaminants.
  • PTFE/silicone construction.
  • Reusable and easy to clean with organic solvents.
  • Compatible with 96- and 384-well plates.
  • Ensures individual well sealing.
  • Maintains sample integrity.