Learn More
MTC Bio Sealing Film Optical Seal Adhesive Polyolefin 2 8mil Ultra-Clear for qPCR 500/Case
Catalog No. 5000460139
Encompass_Preferred
Sealing Film Optical Seal Adhesive Polyolefin 2 8mil Ultra-Clear for qPCR 500/Case
Catalog No. 50-004-60139 Supplier MTC Bio Supplier No. SF070RT9C
Sealing Film Optical Seal Adhesive Polyolefin 2 8mil Ultra-Clear for qPCR 500/Case