Learn More
Thomas Scientific ThermalSeal MiniStrips
SDP
Catalog No. 501458220
Encompass_Preferred
Optimal Performance for Traditional/Classic PCR ThermalSeal Film Designed for basic PCR protocols Withstands temperatures from -40°C to 120°C End tabs facilitate film to plate alignment Cost saving MiniStrip format RNase/DNase/nucleic acid free ThermalSeal a 50 µm polypropylene film with adhesive formulations is optimized for specific PCR applications. Traditional film dimension measures 135.1 x 79.4 mm (123.1 mm without end tabs) and offers sufficient sealing area for most PCR plates. Optimal Performance for Traditional/Classic PCR. ThermalSeal sealing films are not pierceable. For applications where piercing with pipet tips or robotic probes is required for product recovery, see AlumaSeal II sealing films. For real-time PCR applications where maximum optical clarity is required, see ThermalSeal RT and RT2RR sealing films. Note: Not recommended for PCR plates with narrow or irregular sealing surfaces: AlumaSeal, ThermalSeal A™, or ThermalSeal RT&trad
Catalog No. 50-145-8220 Supplier Thomas Scientific Supplier No. 1230Y23
Optimal Performance for Traditional/Classic PCR ThermalSeal Film Designed for basic PCR protocols Withstands temperatures from -40°C to 120°C End tabs facilitate film to plate alignment Cost saving MiniStrip format RNase/DNase/nucleic acid free ThermalSeal a 50 µm polypropylene film with adhesive formulations is optimized for specific PCR applications. Traditional film dimension measures 135.1 x 79.4 mm (123.1 mm without end tabs) and offers sufficient sealing area for most PCR plates. Optimal Performance for Traditional/Classic PCR. ThermalSeal sealing films are not pierceable. For applications where piercing with pipet tips or robotic probes is required for product recovery, see AlumaSeal II sealing films. For real-time PCR applications where maximum optical clarity is required, see ThermalSeal RT and RT2RR sealing films. Note: Not recommended for PCR plates with narrow or irregular sealing surfaces: AlumaSeal, ThermalSeal A™, or ThermalSeal RT&trad