Learn More
Research Products International Corp AeraSeal Sealing Film for Culture Plates, Roll
SDP
Catalog No. 502230227
Encompass
Roll Format for Automation
Adhesive film provides a leak tight seal on culture plates or 96 well assay plates. Unique micro-porous design allows uniform air and CO2 exchange between wells for optimum cell growth or bacterial cultivation. AeraSeal film minimizes cross-contamination between wells and reduces sample evaporation. Film can be easily pierced for sample recovery using a single or multi-channel pipettor.
Medical grade, non-cytotoxic adhesive has a thermal stability range of -20C to 80C.
AeraSeal is a Trademark of Excel Scientific, Inc.
Removable, not reusable.
Catalog No. 50-223-0227 Supplier Research Products International Corp Supplier No. 202504ROLL
Roll Format for Automation
Adhesive film provides a leak tight seal on culture plates or 96 well assay plates. Unique micro-porous design allows uniform air and CO2 exchange between wells for optimum cell growth or bacterial cultivation. AeraSeal film minimizes cross-contamination between wells and reduces sample evaporation. Film can be easily pierced for sample recovery using a single or multi-channel pipettor.
Medical grade, non-cytotoxic adhesive has a thermal stability range of -20C to 80C.
AeraSeal is a Trademark of Excel Scientific, Inc.
Removable, not reusable.