Learn More
Chemglass Life Sciences Chemglass SealPlate Sealing Film, Non-Sterile Polyester, 50 μm, 79.4 x 146.1 mm, For 96-Well Plates, 100/Box (CLS-4001-001)
SDP
Catalog No. NC1937203
Encompass_Preferred
SealPlate sealing film is a 50 μm polyester, non-pierceable film that minimizes evaporation, prevents cross-contamination, and provides a secure seal for microplate wells during storage, incubation, and assays.
  • 50 μm polyester film provides a reliable barrier against evaporation.
  • Dimensions 79.4 x 146.1 mm fit standard 96-well plates.
  • Non-pierceable seal reduces spillage and cross-contamination.
  • Withstands -40° to +120°C for broad assay compatibility.
  • Pack of 100 films for routine laboratory use.
  • Helps reduce edge effects in sensitive plate-based assays.
Catalog No. NC1937203 Supplier Chemglass Life Sciences Supplier No. CLS4001001
SealPlate sealing film is a 50 μm polyester, non-pierceable film that minimizes evaporation, prevents cross-contamination, and provides a secure seal for microplate wells during storage, incubation, and assays.
  • 50 μm polyester film provides a reliable barrier against evaporation.
  • Dimensions 79.4 x 146.1 mm fit standard 96-well plates.
  • Non-pierceable seal reduces spillage and cross-contamination.
  • Withstands -40° to +120°C for broad assay compatibility.
  • Pack of 100 films for routine laboratory use.
  • Helps reduce edge effects in sensitive plate-based assays.